ADVH49B8217C AChemBlock

2,3-Dihydrobenzofuran-4-carbaldehyde

  • Cas Number : 209256-42-8
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-3-dihydrobenzofuran-4-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77098
Availability details for
Quantity
  • Availability
  • Add to Cart