ADVH49B82184 AChemBlock

3-(Boc-amino)-2-piperidinone

  • Cas Number : 99780-98-0
  • Molecular Weight : 214.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-boc-amino-2-piperidinone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70291
Availability details for
Quantity
  • Availability
  • Add to Cart