ADVH49B8218A AChemBlock

imidazo[1,2-a]pyridin-8-ylboronic acid

  • Cas Number : 2058154-31-5
  • Molecular Weight : 161.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/imidazo-1-2-a-pyridin-8-ylboronic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77296
Availability details for
Quantity
  • Availability
  • Add to Cart