ADVH49B8218B AChemBlock

3-bromo-5,5-dimethyl-4H,5H,6H-pyrrolo[1,2-b]pyrazole

  • Cas Number : 2057507-56-7
  • Molecular Weight : 215.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-5-5-dimethyl-4h-5h-6h-pyrrolo-1-2-b-pyrazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77303
Availability details for
Quantity
  • Availability
  • Add to Cart