ADVH49B8218F AChemBlock

tert-butyl (4S)-4-(2-methoxy-2-oxo-ethyl)-2,2-dimethyl-oxazolidine-3-carboxylate

  • Cas Number : 205491-14-1
  • Molecular Weight : 273.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4s-4-2-methoxy-2-oxo-ethyl-2-2-dimethyl-oxazolidine-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77323
Availability details for
Quantity
  • Availability
  • Add to Cart