ADVH49B82193 AChemBlock

2-chloro-6-methyl-thieno[3,2-d]pyrimidine

  • Cas Number : 2033057-21-3
  • Molecular Weight : 184.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-6-methyl-thieno-3-2-d-pyrimidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77359
Availability details for
Quantity
  • Availability
  • Add to Cart