ADVH49B82194 AChemBlock

7-bromo-2-chloro-6-methylthieno[3,2-d]pyrimidine

  • Cas Number : 2033056-28-7
  • Molecular Weight : 263.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-2-chloro-6-methylthieno-3-2-d-pyrimidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77360
Availability details for
Quantity
  • Availability
  • Add to Cart