ADVH49B82195 AChemBlock

tert-butyl 5-(bromomethyl)-3,4-dihydro-1H-isoquinoline-2-carboxylate

  • Cas Number : 2031269-14-2
  • Molecular Weight : 326.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-5-bromomethyl-3-4-dihydro-1h-isoquinoline-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77363
Availability details for
Quantity
  • Availability
  • Add to Cart