ADVH49B8219A AChemBlock

5,5,5-trifluoro-4-methyl-pentan-2-one

  • Cas Number : 200711-56-4
  • Molecular Weight : 154.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-5-5-trifluoro-4-methyl-pentan-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77394
Availability details for
Quantity
  • Availability
  • Add to Cart