ADVH49B8219F AChemBlock

N-Methyl-D-alanine methyl ester hydrochloride

  • Cas Number : 19914-41-1
  • Molecular Weight : 153.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-methyl-d-alanine-methyl-ester-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70358
Availability details for
Quantity
  • Availability
  • Add to Cart