ADVH49B821A1 AChemBlock

tert-butyl N-tert-butoxycarbonyl-N-[(2,6-dichloropyrimidin-4-yl)methyl]carbamate

  • Cas Number : 1984827-51-1
  • Molecular Weight : 378.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-tert-butoxycarbonyl-n-2-6-dichloropyrimidin-4-yl-methyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77440
Availability details for
Quantity
  • Availability
  • Add to Cart