ADVH49B821A2 AChemBlock

5-bromo-6-oxo-1,6-dihydropyridine-3-carbonitrile

  • Cas Number : 19840-44-9
  • Molecular Weight : 199.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-6-oxo-1-6-dihydropyridine-3-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77448
Availability details for
Quantity
  • Availability
  • Add to Cart