ADVH49B821A5 AChemBlock

5-[(4-methoxyphenyl)methoxy]-2-nitro-aniline

  • Cas Number : 1970100-27-6
  • Molecular Weight : 274.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-4-methoxyphenyl-methoxy-2-nitro-aniline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77470
Availability details for
Quantity
  • Availability
  • Add to Cart