ADVH49B821B6 AChemBlock

tert-butyl 2-(methylamino)imidazole-1-carboxylate

  • Cas Number : 1935172-04-5
  • Molecular Weight : 197.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-methylamino-imidazole-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77620
Availability details for
Quantity
  • Availability
  • Add to Cart