ADVH49B821B7 AChemBlock

methyl 2-methyl-1H-pyrrolo[3,2-b]pyridine-3-carboxylate

  • Cas Number : 1934684-80-6
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-methyl-1h-pyrrolo-3-2-b-pyridine-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77634
Availability details for
Quantity
  • Availability
  • Add to Cart