ADVH49B821B8 AChemBlock

tert-butyl N-[(3R,4S)-3-hydroxytetrahydropyran-4-yl]carbamate

  • Cas Number : 1932435-72-7
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-3r-4s-3-hydroxytetrahydropyran-4-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77669
Availability details for
Quantity
  • Availability
  • Add to Cart