ADVH49B821BC AChemBlock

ethyl 4,5,6,7-tetrahydro-1-benzothiophene-2-carboxylate

  • Cas Number : 19282-45-2
  • Molecular Weight : 210.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-5-6-7-tetrahydro-1-benzothiophene-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77707
Availability details for
Quantity
  • Availability
  • Add to Cart