ADVH49B821BE AChemBlock

methyl (1S,2S)-2-(tert-butoxycarbonylamino)cyclopentanecarboxylate

  • Cas Number : 192385-82-3
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-1s-2s-2-tert-butoxycarbonylamino-cyclopentanecarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77714
Availability details for
Quantity
  • Availability
  • Add to Cart