ADVH49B821BF AChemBlock

Tipifarnib S enantiomer

  • Cas Number : 192185-71-0
  • Molecular Weight : 489.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/tipifarnib-s-enantiomer.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86625
Availability details for
Quantity
  • Availability
  • Add to Cart