ADVH49B821C0 AChemBlock

3-nitro-4H-pyrazolo[1,5-a]pyrimidin-5-one

  • Cas Number : 1919868-75-9
  • Molecular Weight : 180.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-nitro-4h-pyrazolo-1-5-a-pyrimidin-5-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77725
Availability details for
Quantity
  • Availability
  • Add to Cart