ADVH49B821C1 AChemBlock

8-methylquinazolin-4(3H)-one

  • Cas Number : 19181-54-5
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/8-methylquinazolin-4-3h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71576
Availability details for
Quantity
  • Availability
  • Add to Cart