ADVH49B821C3 AChemBlock

4-bromo-5-fluoro-2,3-dimethyl-1H-indole-7-carboxamide

  • Cas Number : 1912445-95-4
  • Molecular Weight : 285.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-5-fluoro-2-3-dimethyl-1h-indole-7-carboxamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77749
Availability details for
Quantity
  • Availability
  • Add to Cart