ADVH49B821C6 AChemBlock

(1R,2R)-2-(methoxymethyl)cyclobutanamine;hydrochloride

  • Cas Number : 1909287-01-9
  • Molecular Weight : 151.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-2r-2-methoxymethyl-cyclobutanamine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77761
Availability details for
Quantity
  • Availability
  • Add to Cart