ADVH49B821C7 AChemBlock

BDA-366

  • Cas Number : 1909226-00-1
  • Molecular Weight : 423.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/bda-366.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72366
Availability details for
Quantity
  • Availability
  • Add to Cart