ADVH49B821C8 AChemBlock

tert-butyl N-[(3S,4S)-rel-3-(hydroxymethyl)-4-piperidyl]carbamate

  • Cas Number : 1903840-04-9
  • Molecular Weight : 230.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-3s-4s-rel-3-hydroxymethyl-4-piperidyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77768
Availability details for
Quantity
  • Availability
  • Add to Cart