ADVH49B821C9 AChemBlock

1-benzyl-5,5-dimethylpyrrolidin-3-ol

  • Cas Number : 1903539-91-2
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-benzyl-5-5-dimethylpyrrolidin-3-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77775
Availability details for
Quantity
  • Availability
  • Add to Cart