ADVH49B821CB AChemBlock

2-fluoroterephthalonitrile

  • Cas Number : 1897-53-6
  • Molecular Weight : 146.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-fluoroterephthalonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77788
Availability details for
Quantity
  • Availability
  • Add to Cart