ADVH49B821CD AChemBlock

(3S,4R)-3-fluorooxan-4-amine hydrochloride

  • Cas Number : 1895912-85-2
  • Molecular Weight : 155.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3s-4r-3-fluorooxan-4-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77791
Availability details for
Quantity
  • Availability
  • Add to Cart