ADVH49B821D0 AChemBlock

tert-butyl 7-formyl-3-oxa-9-azabicyclo[3.3.1]nonane-9-carboxylate

  • Cas Number : 1894942-85-8
  • Molecular Weight : 255.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-7-formyl-3-oxa-9-azabicyclo-3-3-1-nonane-9-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77797
Availability details for
Quantity
  • Availability
  • Add to Cart