ADVH49B821D1 AChemBlock

4-(4-bromophenyl)piperidin-2-one

  • Cas Number : 1893051-68-7
  • Molecular Weight : 254.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-4-bromophenyl-piperidin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77800
Availability details for
Quantity
  • Availability
  • Add to Cart