ADVH49B821D3 AChemBlock

1-benzyl 2-methyl (2S,4R)-4-{[(tert-butoxy)carbonyl]amino}pyrrolidine-1,2-dicarboxylate

  • Cas Number : 189215-90-5
  • Molecular Weight : 378.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-benzyl-2-methyl-2s-4r-4-tert-butoxy-carbonyl-amino-pyrrolidine-1-2-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77802
Availability details for
Quantity
  • Availability
  • Add to Cart