ADVH49B821D4 AChemBlock

5-bromo-2-methyl-4-nitro-1H-imidazole

  • Cas Number : 18874-52-7
  • Molecular Weight : 206

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-methyl-4-nitro-1h-imidazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71583
Availability details for
Quantity
  • Availability
  • Add to Cart