ADVH49B821D5 AChemBlock

5-[[(1R)-1-(6-chloro-2-oxo-1H-quinolin-3-yl)ethyl]amino]-1-methyl-6-oxo-pyridine-2-carbonitrile

  • Cas Number : 1887014-14-3
  • Molecular Weight : 354.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-1r-1-6-chloro-2-oxo-1h-quinolin-3-yl-ethyl-amino-1-methyl-6-oxo-pyridine-2-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77821
Availability details for
Quantity
  • Availability
  • Add to Cart