ADVH49B821D7 AChemBlock

3-[(1S)-1-aminoethyl]-6-chloro-1H-quinolin-2-one hydrochloride

  • Cas Number : 1887009-48-4
  • Molecular Weight : 259.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-1s-1-aminoethyl-6-chloro-1h-quinolin-2-one-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77823
Availability details for
Quantity
  • Availability
  • Add to Cart