ADVH49B821D8 AChemBlock

CPI-637

  • Cas Number : 1884712-47-3
  • Molecular Weight : 386.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/cpi-637.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72367
Availability details for
Quantity
  • Availability
  • Add to Cart