ADVH49B821DF AChemBlock

2,2,2-trifluoro-N-(2-mercaptoethyl)acetamide

  • Cas Number : 1869-45-0
  • Molecular Weight : 173.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-2-trifluoro-n-2-mercaptoethyl-acetamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86719
Availability details for
Quantity
  • Availability
  • Add to Cart