ADVH49B821E4 AChemBlock

(3-nitro-1H-pyrazol-5-yl)methanol

  • Cas Number : 1844857-53-9
  • Molecular Weight : 143.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-nitro-1h-pyrazol-5-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S77986
Availability details for
Quantity
  • Availability
  • Add to Cart