ADVH49B821E8 AChemBlock

Sitaxentan

  • Cas Number : 184036-34-8
  • Molecular Weight : 454.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/sitaxentan.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78000
Availability details for
Quantity
  • Availability
  • Add to Cart