ADVH49B821EB AChemBlock

tert-butyl 2-(methoxymethyl)azetidine-1-carboxylate

  • Cas Number : 1824557-53-0
  • Molecular Weight : 201.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-methoxymethyl-azetidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78027
Availability details for
Quantity
  • Availability
  • Add to Cart