ADVH49B821ED AChemBlock

2-Oxa-bicyclo[3.2.0]hept-7-ylamine hydrochloride salt

  • Cas Number : 1824337-15-6
  • Molecular Weight : 149.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-oxa-bicyclo-3-2-0-hept-7-ylamine-hydrochloride-salt.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86653
Availability details for
Quantity
  • Availability
  • Add to Cart