ADVH49B821EF AChemBlock

(5,5-difluorotetrahydropyran-2-yl)methanol

  • Cas Number : 1823910-57-1
  • Molecular Weight : 152.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-5-difluorotetrahydropyran-2-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78057
Availability details for
Quantity
  • Availability
  • Add to Cart