ADVH49B821F3 AChemBlock

ethyl (1R,3S)-3-hydroxycyclohexanecarboxylate

  • Cas Number : 1821824-32-1
  • Molecular Weight : 172.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1r-3s-3-hydroxycyclohexanecarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78132
Availability details for
Quantity
  • Availability
  • Add to Cart