ADVH49B821F4 AChemBlock

(3S,5R)-1-benzyloxycarbonyl-5-methyl-piperidine-3-carboxylic acid

  • Cas Number : 1821737-68-1
  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3s-5r-1-benzyloxycarbonyl-5-methyl-piperidine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78142
Availability details for
Quantity
  • Availability
  • Add to Cart