ADVH49B821F6 AChemBlock

2,5,6-trichloro-3-methyl-3,4-dihydropyrimidin-4-one

  • Cas Number : 1821308-45-5
  • Molecular Weight : 213.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-5-6-trichloro-3-methyl-3-4-dihydropyrimidin-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78147
Availability details for
Quantity
  • Availability
  • Add to Cart