ADVH49B821F7 AChemBlock

5-methyl-1,4,6,7-tetrahydropyrazolo[4,3-c]pyridine dihydrochloride

  • Cas Number : 1820741-75-0
  • Molecular Weight : 210.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-methyl-1-4-6-7-tetrahydropyrazolo-4-3-c-pyridine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78151
Availability details for
Quantity
  • Availability
  • Add to Cart