ADVH49B821FA AChemBlock

(3R,4S)-3-aminotetrahydropyran-4-ol;hydrochloride

  • Cas Number : 1820569-01-4
  • Molecular Weight : 153.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3r-4s-3-aminotetrahydropyran-4-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78157
Availability details for
Quantity
  • Availability
  • Add to Cart