ADVH49B821FC AChemBlock

7-(trifluoromethyl)-3-azabicyclo[4.1.0]heptane hydrochloride

  • Cas Number : 1818847-27-6
  • Molecular Weight : 201.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-trifluoromethyl-3-azabicyclo-4-1-0-heptane-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78160
Availability details for
Quantity
  • Availability
  • Add to Cart