ADVH49B821FE AChemBlock

6-bromo-2-chloroquinoline

  • Cas Number : 1810-71-5
  • Molecular Weight : 242.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-2-chloroquinoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71599
Availability details for
Quantity
  • Availability
  • Add to Cart