ADVH49B82203 AChemBlock

1-(bromomethyl)-4-chloro-2,3-difluorobenzene

  • Cas Number : 1807217-14-6
  • Molecular Weight : 241.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-bromomethyl-4-chloro-2-3-difluorobenzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78199
Availability details for
Quantity
  • Availability
  • Add to Cart