ADVH49B82273 AChemBlock

4-Chloro-3-fluoro-2-methylphenylboronic acid pinacol ester

  • Cas Number : 1680200-54-7
  • Molecular Weight : 288.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-3-fluoro-2-methylphenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70792
Availability details for
Quantity
  • Availability
  • Add to Cart